Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution volumetric solution low in carbonate
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution volumetric solution low in carbonate
Catalog # REGES20101LC
Supplier:  REAGECON
CAS Number:  
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution volumetric solution low in carbonate
Catalog # REGES20101LC
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle GL45
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548