Supplier: VWR Collection
Jump to:
About this item
These bottles are made with 100% virgin HDPE resin.
- Natural colour
- Gamma sterile
Wide neck bottle for easy filling and pouring.
Certifications: Compliance to USFDA and USP Class VI.
Packaging: Individually packed.
7 Options Available Below
Product Details & Documents
These bottles are made with 100% virgin HDPE resin.
- Natural colour
- Gamma sterile
Wide neck bottle for easy filling and pouring.
Certifications: Compliance to USFDA and USP Class VI.
Packaging: Individually packed.
Product Family Options
Product Information
- CapacitéDésignationFiletageHauteurØ ext.AvailabilityPrice
- Specifications:
More
Specifications
Cat. No.216-2933 - Specifications:
More
Specifications
Cat. No.216-2934 - Specifications:
More
Specifications
Cat. No.216-2932 - Specifications:
More
Specifications
Cat. No.216-2935 - Specifications:
More
Specifications
Cat. No.216-2936 - Specifications:
More
Specifications
Cat. No.216-2971 - Specifications:
More
Specifications
Cat. No.216-2931