Order Entry
Belgium
ContactUsLinkComponent
TAAG SP25 Spoilage Wine and Beer Yeast Check SPID
TAAG SP25 Spoilage Wine and Beer Yeast Check SPID
Catalog # TAAGTGG0053/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  
undefined
TAAG SP25 Spoilage Wine and Beer Yeast Check SPID
Catalog # TAAGTGG0053/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  

Specifications

  • Cycler compatibility:
    All real time PCR - Validate for AriaMx and Chai bio
  • Pk:
    12
  • Application:
    RT-qPCR dye
  • Multiplexing:
    Yes
  • One step:
    Yes
  • Description:
    TAAG SP25 spoilage wine and beer yeast check SPID
  • Seq:
    01
  • Contents:
    12 PCR strips 100 µl - 96 reactions

Specifications

About this item

Qualitative detection of spoilage yeast in wine and beer samples. Intended for use in high or low complexity microbiology laboratories.

  • Seamless user experience
  • PCR strips of 8 preloaded with PCR-mix
  • Internal control in each well
  • Reqcuires fam channel
  • Automatic identification done using the TxA app

Can be paired with TAAG X30 extraction kit.

Caution: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.

Extraction Kit

Product Information