TAAG SP24 Spoilage Wine and Beer Bac Check
Catalog # TAAGTGG0052/FROZ
Supplier: KREGLINGER EUROPE
CAS Number:
Specifications
- Cycler compatibility:All real time PCR - Validate for AriaMx and Chai bio
- Pk:12
- Application:RT-qPCR dye
- Multiplexing:Yes
- One step:Yes
- Description:TAAG SP24 spoilage wine and beer bac check
- Seq:01
- Contents:12 PCR strips 100 µl - 96 reactions
Specifications
About this item
Qualitative detection of spoilage bacteria in wine and beer samples. Intended for use in high or low complexity microbiology laboratories.
- Seamless user experience
- PCR strips of 8 preloaded with PCR-mix
- Internal control in each well
- Reqcuires fam channel
- Automatic identification done using the TxA app
Can be paired with TAAG X30 extraction kit.
Caution: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.