TAAG SP22 Spoilage Yeast amd Mold SPID
TAAGTGG0018/FROZ
:
- Cycler compatibility:All real time PCR - Validate for AriaMx and Chai bio
- Model:V-FS02-1
- Pk:12
- Application:RT-qPCR dye
- Multiplexing:Yes
- One step:Yes
- Description:TAAG SP22 spoilage yeast and mold SPID
- Contents:12 PCR strips 100 µl - 96 reactions
Ready-to-use real-time PCR Kit for the detection and identification of over 50 spoilage yeasts and molds.
Can be paired with TAAG E21 Universal Yeast and Mold and TAAG X28 extraction kit.
: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.