Order Entry
Belgium
ContactUsLinkComponent
TAAG SP12 Spoilage Bacteria SPID
TAAG SP12 Spoilage Bacteria SPID
Catalog # TAAGTGG0017/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  
TAAG SP12 Spoilage Bacteria SPID
Catalog # TAAGTGG0017/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  

Specifications

  • Cycler compatibility:
    All real time PCR - Validate for AriaMx and Chai bio
  • Model:
    V-FS01-1
  • Pk:
    12
  • Application:
    RT-qPCR dye
  • Multiplexing:
    Yes
  • One step:
    Yes
  • Description:
    TAAG SP12 spoilage bacteria SPID
  • Contents:
    12 PCR strips 100 µl - 96 reactions

Specifications

About this item

Ready-to-use real-time PCR Kit for the detection and identification of over 50 spoilage bacteria.

  • Seamless user experience
  • PCR strips of 8 preloaded with PCR-mix
  • Internal control in each well
  • Reqcuires fam channel
  • Automatic identification done using the TxA app

Can be paried with TAAG E11 Universal Bacteria growth medium and TAAG X29 extraction kit.

Caution: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.