TAAG F13 SPID
Catalog # TAAGTGG0049/FROZ
Supplier: KREGLINGER EUROPE
CAS Number:
Specifications
- Cycler compatibility:All real time PCR - Validate for AriaMx and Chai bio
- Model:V-FF16-2
- Pk:12
- Application:RT-qPCR dye
- Multiplexing:Yes
- One step:Yes
- Description:Ready to use multiplex real-time PCR kit
- Seq:01
- Contents:12 PCR strips 100 µl - 96 reactions
Specifications
About this item
Ready-to-use multiplex real-time PCR kit for the simultaneous detection and identification of E. coli from enriched samples using TAAG E41 pathogens food or TAAG E31 pathogens surfaces.
Caution: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.