TAAG F23 SPID
TAAGTGG0014/FROZ
:
- Cycler compatibility:All real time PCR - Validate for AriaMx and Chai bio
- Model:V-FF12-2
- Pk:12
- Application:RT-qPCR dye
- Multiplexing:Yes
- One step:Yes
- Description:TAAG F23 Duplex Listeria SPID
- Seq:01
- Contents:12 PCR strips 100 µl - 96 reactions
Ready-to-use multiplex real-time PCR kit for the simultaneous detection and identification of Listeria spp and Listeria monocytogenes.
- Seamless user experience
- PCR strips of 8 preloaded with PCR-mix
- Internal control in each well
- Reqcuires fam channel
Can be paired with the selective growth medium TAAG E13 universal Listera used for enriching food and beverage samples or TAAG E31 pathogens surfaces, growth medium suitable for surface samples colleted using the neutrosampling S11.
Easy DNA extraction usign the TAAG X29 kit.
Automatic result interpretation by TxA app.
: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.