Order Entry
Belgium
ContactUsLinkComponent
TAAG F25 SPID
TAAG F25 SPID
Catalog # TAAGTGG0016/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  
TAAG F25 SPID
Catalog # TAAGTGG0016/FROZ
Supplier:  KREGLINGER EUROPE
CAS Number:  

Specifications

  • Cycler compatibility:
    All real time PCR - Validate for AriaMx and Chai bio
  • Model:
    V-FF22-2
  • Pk:
    12
  • Application:
    RT-qPCR dye
  • Inhibitor-rich sample:
    Yes
  • Multiplexing:
    Yes
  • One step:
    Yes
  • Description:
    TAAG F25 Duplex Salm Lmon SPID
  • Seq:
    01
  • Contents:
    12 PCR strips 100 µl - 96 reactions

Specifications

About this item

Ready-to-use multiplex real-time PCR kit for the simultaneous detection and identification of Listeria monocytogenes and Salmonella.

  • Seamless user experience
  • PCR strips of 8 preloaded with PCR-mix
  • Internal control in each well
  • Reqcuires fam channel

Can be paired with the selective growth medium TAAG E41 pathogens used for enriching food and beverage samples or TAAG E31 pathogens surfaces, growth medium suitable for surface samples colleted using the neutrosampling S11.

Easy DNA extraction usign the TAAG X29 kit.

Automatic result interpretation by TxA app.

Caution: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.

Sample Collection

Product Information