ContactUsLinkComponent
TAAG E31 Universal Surfaces
TAAG E31 Universal Surfaces
Catalog # TAAGTGG0002
Supplier:  KREGLINGER EUROPE
CAS Number:  
undefined
TAAG E31 Universal Surfaces
Catalog # TAAGTGG0002
Supplier:  KREGLINGER EUROPE
CAS Number:  
Specifications
  • Model:
    V-FF03-1
  • Pk:
    1
  • Description:
    TAAG E31 universal surfaces
  • Contents:
    1 jar - 500 g
Specifications
About this item
Dehydrated enrichment media for growing Salmonella spp., Staphylococcus aureus, Escherichia coli and Listeria monocytogenes in surface samples. It contains selected growth factors and nutritional requirements optimized to promote pathogens development.