ContactUsLinkComponent
Nitrile gloves, Kimtech™ Polaris™ Xtra
Nitrile gloves, Kimtech™ Polaris™ Xtra
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Certificates
undefined
Nitrile gloves, Kimtech™ Polaris™ Xtra
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Specifications
  • Catalog No:
  • 112-6565
  • 112-6566
  • 112-6567
  • 112-6568
  • 112-6569
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,11 mil
  • 0,11 mil
  • 0,11 mil
  • 0,11 mil
  • 0,11 mil
  • Finger thickness:
  • 0,19 mil
  • 0,19 mil
  • 0,19 mil
  • 0,19 mil
  • 0,19 mil
  • Palm thickness:
  • 0,14 mil
  • 0,14 mil
  • 0,14 mil
  • 0,14 mil
  • 0,14 mil
  • FDA regulated:
  • +
  • +
  • +
  • +
  • +
  • Tensile strength:
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • Ultimate elongation:
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • EN norm:
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • ASTM No.:
  • D6978
  • D6978
  • D6978
  • D6978
  • D6978