ContactUsLinkComponent
Kimtech™ Polaris™ Nitrile Gloves
undefined
Kimtech™ Polaris™ Nitrile Gloves
  • Catalog No:
  • 112-6562
  • 112-6563
  • 112-6560
  • 112-6564
  • 112-6561
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,09 mil
  • 0,09 mil
  • 0,09 mil
  • 0,09 mil
  • 0,09 mil
  • Finger thickness:
  • 0,15 mil
  • 0,15 mil
  • 0,15 mil
  • 0,15 mil
  • 0,15 mil
  • Palm thickness:
  • 0,12 mil
  • 0,12 mil
  • 0,12 mil
  • 0,12 mil
  • 0,12 mil
  • FDA regulated:
  • +
  • +
  • +
  • +
  • +
  • Tensile strength:
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • ≥ 14 Mpa
  • Ultimate elongation:
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • ≥ 400%, min.
  • EN norm:
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • EN ISO 374-1 Type B (JKOPT)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • ASTM No.:
  • D6978
  • D6978
  • D6978
  • D6978
  • D6978
Frequently Bought Together