ContactUsLinkComponent
Notrax® 562 Sanitop® Floor Mattings
Notrax® 562 Sanitop® Floor Mattings
Catalog # JUSU562S0035BL
Notrax® 562 Sanitop® Floor Mattings
Catalog # JUSU562S0035BL
Specifications
  • Colour:
    Black
  • Description:
    Rubber mat
  • L×W:
    1520×910 mm
  • Thickness:
    13 mm (½")
  • Pk:
    1
Specifications
About this item
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
  • Durable rubber compound
  • Molded alternating large/small hole drainage system keeps moisture and debris free from work area
  • Raised pattern surrounds large holes to channel drainage and provide worker traction
  • Beveled ramps on all 4 sides
  • 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards. Sanitop® offers a simple, economical solution for wet environments in many industrial or food service applications.
Weight: 9,97903 kg