

Cyclohexylamine solution 0.1 mol/l (0.1 M) in 2-propanol
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:108-91-8
:
Formula:C₆H₁₃N
MDL Number:MFCD00001486
:99.18 g/mol
Density:0,7865 g/cm³
Flash Pt:<21 °C
UN:1993
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together