Order Entry
Belgium
ContactUsLinkComponent
Acetone ≥99.8% VLSI, Electronic Grade
Inflammable
Irritant
Acetone ≥99.8% VLSI, Electronic Grade
N° de catalogue VWRC6683.2500
Fournisseur:  VWR Chemicals
Numéro CAS:  
undefined
Inflammable
Irritant
Acetone ≥99.8% VLSI, Electronic Grade
N° de catalogue VWRC6683.2500
Fournisseur:  VWR Chemicals
Numéro CAS:  

Spécifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Spécifications

Résultats des tests de spécification

  • Assay (GC):
    ≥ 99.8 %
  • Identification (IR):
    Passes test
  • Colouration:
    ≤ 10 APHA
  • Particle size (> 0.50 μm) (/ml) (*):
    ≤ 200
  • Resistivity (MOhm-cm):
    ≥ 5
  • Aldehyde (as HCHO):
    ≤ 10 ppm
  • Ethanol (GC):
    ≤ 100 ppm
  • Evaporation residue:
    ≤ 3 ppm
  • Free acid (as CH₃COOH):
    ≤ 20 ppm
  • Free alkali (as NH₃):
    ≤ 2 ppm
  • Methanol (GC):
    ≤ 0.05 %
  • Substances reducing KMnO₄ (as O):
    ≤ 2.5 ppm
  • Water:
    ≤ 0.2 %
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 50 ppb
  • As (Arsenic):
    ≤ 10 ppb
  • Au (Gold):
    ≤ 20 ppb
  • B (Boron):
    ≤ 10 ppb
  • Ba (Barium):
    ≤ 20 ppb
  • Be (Beryllium):
    ≤ 10 ppb
  • Bi (Bismuth):
    ≤ 20 ppb
  • Ca (Calcium):
    ≤ 100 ppb
  • Cd (Cadmium):
    ≤ 10 ppb
  • Co (Cobalt):
    ≤ 10 ppb
  • Cr (Chromium):
    ≤ 10 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 10 ppb
  • Ga (Gallium):
    ≤ 10 ppb
  • In (Indium):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 20 ppb
  • Li (Lithium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 20 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Mo (Molybdenum):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 100 ppb
  • Ni (Nickel):
    ≤ 10 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Pt (Platinum):
    ≤ 50 ppb
  • Sb (Antimony):
    ≤ 10 ppb
  • Sn (Tin):
    ≤ 20 ppb
  • Sr (Strontium):
    ≤ 10 ppb
  • Ti (Titanium):
    ≤ 20 ppb
  • Tl (Thallium):
    ≤ 10 ppb
  • V (Vanadium):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 20 ppb
  • Zr (Zirconium):
    ≤ 20 ppb
  • Additional information:
    (*): Particle size (> 0.50 μm) (/ml) in raw material before filling

Résultats des tests de spécification

À propos de cet article

 :
 :58.08 g/mol
Index Merck:13,00067
Point d'ébullition:56.2 °C (1013 hPa)
Point de fusion:–95.4 °C
Densité:0.792 g/cm³ (20 °C)
Point d'éclair:<–20 °C (closed cup)
Température de stockage:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
Numéro CAS:67-64-1
Formule:CH₃COCH₃
Numéro MDL:MFCD00008765