Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 36% VLSI, Electronic Grade
 
 
Hydrochloric acid 36% VLSI, Electronic Grade
  50489136.320
 :  
undefined
 
 
Hydrochloric acid 36% VLSI, Electronic Grade
  50489136.320
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay (acidimetric):
    36.0 - 38.0 %
  • Colouration:
    ≤ 10 APHA
  • Particle size(>0.50 μm)(/ml)(at filling):
    ≤ 64
  • Free halogen (as Cl₂):
    ≤ 0.5 ppm
  • Non-volatile matter:
    ≤ 5
  • NH₄ (Ammonium):
    ≤ 1000 ppb
  • PO₄ (Phosphate):
    ≤ 100 ppb
  • SO₃ (Sulphite):
    ≤ 500 ppb
  • SO₄ (Sulphate):
    ≤ 500 ppb
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 50 ppb
  • As (Arsenic):
    ≤ 5 ppb
  • Au (Gold):
    ≤ 20 ppb
  • B (Boron):
    ≤ 10 ppb
  • Ba (Barium):
    ≤ 10 ppb
  • Be (Beryllium):
    ≤ 10 ppb
  • Bi (Bismuth):
    ≤ 20 ppb
  • Ca (Calcium):
    ≤ 200 ppb
  • Cd (Cadmium):
    ≤ 5 ppb
  • Co (Cobalt):
    ≤ 10 ppb
  • Cr (Chromium):
    ≤ 10 ppb
  • Cu (Copper):
    ≤ 10 ppb
  • Fe (Iron):
    ≤ 100 ppb
  • Ga (Gallium):
    ≤ 20 ppb
  • Ge (Germanium):
    ≤ 50 ppb
  • In (Indium):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 50 ppb
  • Li (Lithium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 50 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Mo (Molybdenum):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 50 ppb
  • Ni (Nickel):
    ≤ 10 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Pt (Platinum):
    ≤ 20 ppb
  • Sn (Tin):
    ≤ 20 ppb
  • Sr (Strontium):
    ≤ 10 ppb
  • Ti (Titanium):
    ≤ 10 ppb
  • Tl (Thallium):
    ≤ 20 ppb
  • V (Vanadium):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 50 ppb
  • Zr (Zirconium):
    ≤ 10 ppb

 

 

 :
 :110 °C (1013 hPa)
 :–30 °C
 :1.18 g/cm³ (20 °C)
 :8,II
 :36.46 g/mol
 :Ambient
 :231-595-7
 :1789
 :01-2119484862-27
 :GHS05
 :GHS07
 :7647-01-0
 :HCl
 :MFCD00011324