Orders LinkContactUsLinkComponent
HIGH STAT PLUS ESD Labcoats
Supplier:   ALSICO HIGHTECH
HIGH STAT PLUS ESD Labcoats
Supplier:  ALSICO HIGHTECH
Certificates
HIGH STAT PLUS ESD Labcoats
15 Options Available Below
Product Details & Documents
Unisex ESD labcoat with long sleeves.
  • Reverse collar
  • Centre front closure with hidden press studs
  • 1 chest pocket on the left
  • 1 inside pocket at chest level on the right
  • 2 patched side pockets
  • Inset sleeves, adjustable with press studs at the bottom
  • ESD logo
Industrial launderable.
 : Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.
Specifications for Product Family
Specifications
  • Catalog No:
  • ALSO1000709-XS
  • ALSW1031122-S
  • ALSO1000709-2XL
  • ALSO1002183-XS
  • ALSO1002183-S
  • ALSO1002183-M
  • ALSO1002183-L
  • ALSO1002183-XL
  • ALSO1002183-2XL
  • ALSO1000547-XS
  • ALSO1000547-S
  • ALSO1000547-M
  • ALSO1000547-L
  • ALSO1000547-XL
  • ALSO1000547-2XL
  • Garment application:
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Cleanroom|Industrial
  • Risk area:
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • Weight:
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
Specifications
Product Family Options
Product Information
  • Type
    Colour
    Clothing Size
    Garment fit
    Availability
    Price
  • ESD
    White
    XS
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,20 €
    69,20 €
    1 * 1 ST
       69,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000709-XS
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    White
    S
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,50 €
    56,50 €
    1 * 1 ST
       56,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1031122-S
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    White
    XXL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,20 €
    69,20 €
    1 * 1 ST
       69,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000709-2XL
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    XS
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-XS
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    S
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-S
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    M
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-M
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    L
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-L
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    XL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-XL
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Grey
    XXL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1002183-2XL
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    XS
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-XS
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    S
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-S
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    M
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-M
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    L
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-L
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    XL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-XL
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²
  • ESD
    Blue
    XXL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,30 €
    77,30 €
    1 * 1 ST
       77,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1000547-2XL
    Garment application
    Cleanroom|Industrial
    Risk area
    ESD
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    68% polyester/30,5% cotton/1,5% carbon
    Weight
    175 g/m²