Orders LinkContactUsLinkComponent
APC 2 FLASH Multirisk Labcoats
APC 2 FLASH Multirisk Labcoats
Supplier:   ALSICO
APC 2 FLASH Multirisk Labcoats
Supplier:  ALSICO
Certificates
APC 2 FLASH Multirisk Labcoats
7 Options Available Below
Product Details & Documents
Multirisk labcoat especially designed for APC 2.
  • ATPV value: 26,6 cal/cm²
  • ELIM value: 21 cal/cm²
  • Modern fit
  • Upright, rounded collar
  • Centre front closure with hidden press studs, ECO design
  • Inside zipped pocket
  • 2 large side pockets with flap and hidden press studs
  • Long sleeves with sleeve cuffs and elastic
  • Piping in accent colour
Industrial launderable.
Certifications: Oeko-Tex® standard 100; EN 1149-5 2018; IEC 61482 - 2: 2018 - APC2; EN ISO 11612:2015 (A1, B1, C1, D0, E0, F1); EN 11611:2015 - Class 2 A1; EN 13034 +A1.
Specifications for Product Family
Specifications
  • Catalog No:
  • ALSW1030929-S
  • ALSW1030929-M
  • ALSW1030929-L
  • ALSW1030929-XL
  • ALSW1030929-2XL
  • ALSW1030929-3XL
  • ALSW1030929-4XL
  • Garment application:
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
  • Risk area:
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Flame-retardant|Anti-static
  • Gender:
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Fabric:
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
  • Weight:
  • 480 g/m²
  • 480 g/m²
  • 480 g/m²
  • 480 g/m²
  • 480 g/m²
  • 480 g/m²
  • 480 g/m²
Specifications
Product Family Options
Product Information
  • Type
    Colour
    Clothing Size
    Garment fit
    Availability
    Price
  • Multirisk
    Grey
    S
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    1 * 1 ST
       217,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-S
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    M
    Regular
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    Log in for availability
    Each
    Each - 226,00 €
    226,00 €
    1 * 1 ST
       217,00 €
    Each
       226,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-M
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    L
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    1 * 1 ST
       217,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-L
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    XL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    1 * 1 ST
       217,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-XL
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    XXL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    1 * 1 ST
       217,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-2XL
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    3XL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-3XL
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²
  • Multirisk
    Grey
    4XL
    Regular
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSW1030929-4XL
    Garment application
    Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
    Risk area
    Flame-retardant|Anti-static
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    Outer fabric: 54% modacrylic/44% cotton/2% antistatic
    Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
    Weight
    480 g/m²