Antistatic Shell Gloves, A197
Supplier: Portwest
Certificates
About this item
Constructed from an advanced polyester and carbon fibre blend. This glove protects equipment like circuit boards from the natural oils of the skin while also protecting sensitive equipment from harmful static discharge.
- Suitable for use in ESD environments
- This glove can be used with most mobile touchscreen devices
- Seamless 13 gauge liner
Low linting construction for minimal contamination. Retail tag aids presentation for retail sales.
Colour: Grey
Certifications: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 113XX, EN 16350:2014 Pass Level R <1.0 x 108 Ω -. IEC 61340-5-1:2016. CE certified.
Specifications
- Catalog No:
- PRTWA197GRRXL
- PRTWA197GRRS
- PRTWA197GRRM
- PRTWA197GRRL
- PRTWA197GRRXS
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Polyester/carbon fibre
- Polyester/carbon fibre
- Polyester/carbon fibre
- Polyester/carbon fibre
- Polyester/carbon fibre
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile