Orders LinkContactUsLinkComponent
Polka Dot Plus Gloves
Polka Dot Plus Gloves
Catalog #
Polka Dot Plus Gloves
Catalog #
Supplier:  
Polka Dot Plus Gloves
Polka Dot Plus Gloves
Product Details & Documents
PVC dots on both sides extend the life of this glove. Perfect for warehousing and distribution, these ambidextrous gloves can be worn on either hand.
  • PVC dotted palm for enhanced grip
  • Ambidextrous design suitable for left- and right-handed users
  • Seamless 13 gauge liner
  • Performs well in dry conditions
Low linting construction for minimal contamination, with a breathable seamless liner.
Colour: White/blue
Certifications: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 1142X. CE certified.