Order Entry
Belgium
Orders LinkContactUsLinkComponent
ES Buffer

ES Buffer

ES-F buffer, 6 vials×6,0 ml
Catalog # WAKFESB-0006
ES Buffer
Catalog # WAKFESB-0006
CAS Number:  
ES Buffer
ES Buffer

ES-F buffer, 6 vials×6,0 ml

CAS Number:  
Price
367,00 €
Log in for availability
1 * 36 mL Each
Product Details & Documents

The ES Buffer is an endotoxin-specific buffer that is ideal for untreated LAL.

  • Each vial has solution for use in reconstituting a max of 6,0 ml of reagent
  • Ideal for use in reconstituting any LAL that has the potential to react with glucans, thus making an endotoxin-specific reagent

Though all LAL reagents produced by FUJIFILM Wako LAL are endotoxin-specific, other commercially available LAL reagents are not. Without this buffer, tests can present a potential false positive when trying to determine the endotoxin concentration in the sample.

The ES Buffer works by adding a large amount of carboxymethylated curdlan (CMC) to LAL, thus preventing the activation of LAL by glucan. The presence of large amounts of glucan does not interfere with the quantitation of endotoxin. FUJIFILM Wako first made use of these findings by developing this endotoxin-specific buffer, which contains high concentrations of CMC. When the ES Buffer is used to reconstitute LAL, reagent becomes endotoxin-specific.

Documents

Specifications

Specifications

  • Description:
    ES-F buffer, 6 vials×6,0 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.