Order Entry
Belgium
ContactUsLinkComponent
Accessories for Endotoxin Tests
Accessories for Endotoxin Tests
Catalog # WAKF293-51601
CAS Number:  
undefined
Accessories for Endotoxin Tests
Catalog # WAKF293-51601
CAS Number:  

Specifications

  • Pk:
    40 mL
  • Description:
    Endotoxin Extracting Solution for LAL Test

Specifications

About this item

This endotoxin-free solution containing human serum albumin (HSA) is capable of extracting endotoxins that cannot be extracted in water or saline solution.

Traditionally, water or saline solution has been used to extract endotoxin in tests involving medical devices and equipment; however, the efficacy of this extraction method is debated. By using this extraction solution, it provides our customers with a more reliable method for medical device testing.

Recommended for use in endotoxin testing on equipment and devices that may come into contact with fluids containing blood or protein.