Order Entry
Belgium
ContactUsLinkComponent
HTD Buffer Exchange and Desalting Device, HTDialysis
HTD Buffer Exchange and Desalting Device, HTDialysis
Catalog # HTDL1011
Supplier:  HTDIALYSIS
undefined
HTD Buffer Exchange and Desalting Device, HTDialysis
Catalog # HTDL1011
Supplier:  HTDIALYSIS

Specifications

  • Volume:
    0,050 - 0,150 ml
  • Description:
    12 well buffer exchange and desalting system, time to equilibrium 2 - 4 hours
  • Pk:
    1

Specifications

About this item

HTD 12A reusable 12 well buffer exchange and desalting device. 50 to 150 µl capacity. Sample volume dialysate ration of 1:26. Compatible with a wide range of dialysis membrane pore sizes.

  • 1:26 ratio of sample volume to dialysate
  • Minimal non-specific binding due to 100% Teflon block construction
  • Ready in minutes and achieves equilibrium in 2 to 4 hours on average
  • Conserves valuable reagents with working volumes of 30 to 150 µl
  • Automatable with standard robotic workstations and 96-well pipettors
  • Ready access to both the sample and dialysate sides of the membrane from the top of the device
  • Compatible with a wide range of dialysis membrane pore sizes i.e. 1 kDa, 3,5 kDa, 6 to 8 kDa, 10 kDa, 12 to 14 kDa, 25 kDa, and 50 kDa
  • Teflon block compatible with all HTD96 universal base

The reusable HTD12a device was developed to automate low volume buffer exchange and desalting applications. The device was designed with a large sample to dialysate ratio to promote rapid desalting or buffer exchange.

Applications:
- All desalting and buffer exchange steps of sample processing applications: dialysis of proteins, oligonucleotides, RNA or DNA samples
- Removal of sample contaminates - salts, solvents, detergents
- Conducting Formation studies - protein-protein, protein-DNA, protein-RNA

Additional specifications:
Dimensions: 170,1×116,8×38,1 mm
Weight: 0,5 kg

Membranes

Product Information