Orders LinkContactUsLinkComponent
Software for Evolution™ UV/VIS Spectrophotometers
Software for Evolution™ UV/VIS Spectrophotometers
Software for Evolution™ UV/VIS Spectrophotometers
Certificates
Software for Evolution™ UV/VIS Spectrophotometers
Software for Evolution™ UV/VIS Spectrophotometers
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • SPET837-004000
    Insight Pro software with security, defines workflow with data integrity, includes Insight™ Pro software with compliance with the FDA 21 CFR Part 11 regulations
    Insight Pro software with security, defines workflow with data integrity, includes Insight™ Pro software with compliance with the FDA 21 CFR Part 11 regulations
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 150,00 €
    4 150,00 €
    1 * 1 ST
       4 150,00 €
  • SPET837-003900
    Insight Pro software, workflow definition, includes software for Evolution™ spectrophotometers with preprogrammed Life science methods
    Insight Pro software, workflow definition, includes software for Evolution™ spectrophotometers with preprogrammed Life science methods
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • SPET869-175600
    Visionlite colorcalc, to measure color, includes software to perform color calculations from full spectrum scan of Evolution spectrophotometers
    Visionlite colorcalc, to measure color, includes software to perform color calculations from full spectrum scan of Evolution spectrophotometers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 390,00 €
    4 390,00 €
    1 * 1 ST
       4 390,00 €