Manual Vial Crimping and Decapping Tongs
A range of high-quality tongs and pliers for the crimping and decapping of pharmaceutical vials.
- Options for 'plain', 'flip top/flip off', and 'flip tear up' seals
- Crimping and decapping of 13 and 20 mm seals
- Fast and reliable, for a professional quality finish
- Easy, intuitive operation
All devices feature hardened, durable steel jaws to ensure reproducibility of seal crimp quality. They also feature an adjustable stop to ensure that equal pressure is applied to each crimp. Decapping pliers use a press and wrench action for removing seals from vials. The handles are plastic-coated for operator comfort.
Adelphi manufacturing also offers the semi-automatic powered vial crimper, to reduce operator fatigue.
Frequently Bought Together