ContactUsLinkComponent
Density Meters with Peltier Temperature Control, DSG
Density Meters with Peltier Temperature Control, DSG
Supplier:  BELLINGHAM STANLEY
Certificates
undefined
Density Meters with Peltier Temperature Control, DSG
Supplier:  BELLINGHAM STANLEY
Specifications
  • Catalog No:
  • 614-0345
  • 614-0346
  • Model:
  • DSG50
  • DSG40
  • Density resolution:
  • 0,00001 g/cm³
  • 0,0001 g/cm³
  • Density accuracy:
  • ±0,00005 g/cm³
  • ±0,0001 g/cm³
  • Reproducibility:
  • ±0,00002 g/cm³
  • ±0,0001 g/cm³
  • Measuring range:
  • 0 - 3 g/cm³
  • 0 - 3 g/cm³
  • Temperature accuracy:
  • ±0,03 °C
  • ±0,05 °C
  • Temperature range:
  • 10…95 °C
  • 10…95 °C