À propos de cet article
Thermo Scientific™ TSX Series high-performance laboratory refrigerators offer a sustainable choice for clinical and laboratory refrigeration.
- Designed to safely store medical and laboratory storage
- Whisper quiet with a sound level of 52 dBA
- V-drive technology
- Up to 40% less energy usage than conventional refrigerant models
- Choice of glass or solid door models
- Lower HVAC costs as a result of less heat emission into the environment
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3 to 7 °C.
Designed to safely store medical and laboratory storage, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2 to 8 °C. Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. These refrigerators offer up to 40% less energy usage than conventional refrigerant models.
Refrigerators feature an automatic defrost and a microprocessor display.
Manufactured in an award-winning, zero-waste to landfill facility. Operating temperature range of 2 to 8 °C, factory setpoint at 5 °C.
Note: Dimensions shown below are W×D×H.
Spécifications
- Catalog No:
- HERATSX3005GV
- HERATSX1205SV
- HERATSX1205GV
- HERATSX3005SV
- HERATSX5005GV
- HERATSX5005SV
- HERATSX4505GV
- HERATSX4505GA
- Model:
- TSX3005GV
- TSX1205SV
- TSX1205GV
- TSX3005SV
- TSX5005GV
- TSX5005SV
- TSX4505GV
- TSX4505GA
- Display:
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Defrost:
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Shelves/drawers:
- 4 full size shelves
- 4 full size shelves
- 4 full size shelves
- 4 full size shelves
- 8 full size shelves
- 8 full size shelves
- 8 full size shelves
- 8 full size shelves
- Door type:
- 1 glass swing door
- 1 solid swing door
- 1 glass swing door
- 1 solid swing door
- 2 double glass door
- 2 double solid door
- 2 glass sliding door
- 1 glass sliding door
- Refrigerant:
- R290
- R290
- R290
- R290
- R290
- R290
- R290
- R290
- Electrical:
- 230 V, 50 Hz, 15 A
- 230 V, 50 Hz, 15 A
- 230 V, 50 Hz, 15 A
- 230 V, 50 Hz, 15 A
- 115 V, 60 Hz, 20 A
- 208 - 230 V, 50 Hz, 15 A
- 115 V, 60 Hz, 20 A
- 115 V, 60 Hz, 20 A
- Weight:
- 177 kg
- 130 kg
- 129 kg
- 177 kg
- 303 kg
- 303 kg
- 277 kg
- 277 kg