Fit Test Adapters
Fit test adapter, quantitative, 601
Catalog # MMMA601
Supplier: 3M
Jump to:
Product Details & Documents
For an alternative method to sharing a probed fit testing respirator, use the 3M™ quantitative fit test adapter. The adaptor kits allow users to fit test with a personal 3M™ Respirator, rather than using a shared probed fit test respirator.
- Adapter allows fit testing with an individual's personal 3M™ respirator
- Alternative to sharing a probed fit testing respirator
- Quick and easy to fit
The 3M™ Quantitative fit test adapters are designed for use with 3M™ half and full-face respirators with bayonet-style filter attachments.
Specifications
- Description:Fit test adapter, quantitative, 601