Orders LinkContactUsLinkComponent
Elevated Temperature Front Helmet Cover
Elevated Temperature Front Helmet Cover
Elevated temperature front cover, aluminum, Silver
Catalog # MMMAFC1-AL
Supplier:   3M
Elevated Temperature Front Helmet Cover
Catalog # MMMAFC1-AL
Supplier:  3M
Elevated Temperature Front Helmet Cover
Elevated Temperature Front Helmet Cover
Elevated temperature front cover, aluminum, Silver
Price
114,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The elevated temperature front cover is a silver aluminium front cover designed to fit on to 3M™ hard hats.
  • Provide additional protection from sparks and metal splatter
  • Streamlined design prevents interference with other equipment during use
  • Fabric parts can be easily replaced when needed
Get extra protection from sparks and metal splatter on the front of a hard hat with the elevated temperature front cover. This silver aluminium hard hat cover has a streamlined design that helps prevent interference with other equipment during use. It fits on to 3M™ hard hats for added protection and to help reduce the possibility of particulate dropping into the users face when a faceshield is lifted. The fabric parts can be easily replaced when needed.
Specifications
  • Colour:
    Silver
  • Description:
    Elevated temperature front cover, aluminum