Adflo™ Particulate and Gas Filters
Supplier: 3M
Certificates
About this item
Adflo™ particulate and gas filters are replacement, high-efficiency filters designed for use with 3M™ Adflo™ powered air purifying respirators.
- Provides protection against a range of particulate and gas hazards (dependent upon filters selected)
- For use with 3M™ Adflo™ Powered Air Purifying Respirators (PAPR)
- Particle filter, gas filters, and odour filter/pad available
- Installation is easy and secure
- For highest possible safety and comfort, regularly check equipment for worn or damaged parts
Get protection from a range of particulate and gas hazards with Adflo™ particulate and gas filters. These filters, compatible with the 3M™ Adflo™ powered air respirator system is simple to use, easy to install, and they provide high efficiency protection. The design allows users to replace the particulate or gas filter independently, and there is no need to replace both at the same time.
Frequently Bought Together