ContactUsLinkComponent
Cole-Parmer® HM-100 Uncontrolled Heating Mantles
Cole-Parmer® HM-100 Uncontrolled Heating Mantles
Supplier:  Cole-Parmer
Certificates
undefined
Cole-Parmer® HM-100 Uncontrolled Heating Mantles
Supplier:  Cole-Parmer
Specifications
  • Catalog No:
  • ELET36003-06
  • ELET36003-14
  • ELET36103-05
  • ELET36003-18
  • Electrical:
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • Max. temperature:
  • 450 °C
  • 450 °C
  • 450 °C
  • 450 °C