ContactUsLinkComponent
HAEMAFlex™ 6/8 Swinging Bucket Rotors
HAEMAFlex™ 6/8 Swinging Bucket Rotors
Fournisseur:  Thermo Scientific
Certificats
undefined
HAEMAFlex™ 6/8 Swinging Bucket Rotors
Fournisseur:  Thermo Scientific
Spécifications
  • Catalog No:
  • HERA75003861
  • HERA75003881
  • Capacity:
  • 6×550 ml blood bags (6×1000 ml)
  • 8×550 ml blood bags (8×1000 ml)
  • Rotor type:
  • Swinging bucket rotor
  • Swinging bucket rotor
  • Max. RCF:
  • 7295×g
  • 7144×g
  • Max. speed:
  • 5000 min⁻¹
  • 4600 min⁻¹
Spécifications
Frequently Bought Together