Orders LinkContactUsLinkComponent
Plastic Box, Translucent Polypropylene
Supplier:   VWR Collection
Plastic Box, Translucent Polypropylene
Supplier:  VWR Collection
Certificates
Plastic Box, Translucent Polypropylene
Plastic Box, Translucent Polypropylene
12 Options Available Below
Product Details & Documents
This range of plastic boxes offers a modern and practical solution for the storage, organization, transportation, and presentation of small parts. With their innovative design, these plastic boxes are ideal for various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
  • 5 available formats: A4, A5, A6, A7, and A8
  • Material: 100% recyclable virgin polypropylene
  • Hinged lid with secure snap closure
  • Excellent stackability due to stacking feet
  • Temperature resistance from –20 to +80 °C
  • Clear lid for excellent content visibility
  • Dividers reaching the top of the box to prevent movement of small items between compartments
  • Fixed and removable compartments for A4, A5, and A6 formats, fixed compartments for A7 and A8 formats
Their versatility makes them ideal for many applications in various sectors such as optics, electronics, laboratories, industry, museums, and the medical field.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 216-3024
    Plastic box, 1 compartment, compartment size 82×53 mm
    Plastic box, 1 compartment, compartment size 82×53 mm
    90×60×20 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 47,80 €
    47,80 €
    1 * 24 ST
       47,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3024
  • 216-3025
    Plastic box, 2 compartments, compartment size 53×40 mm
    Plastic box, 2 compartments, compartment size 53×40 mm
    90×60×20 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 54,10 €
    54,10 €
    1 * 24 ST
       54,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3025
  • 216-3026
    Plastic box, 4 compartments, compartment size 40×26 mm
    Plastic box, 4 compartments, compartment size 40×26 mm
    90×60×20 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 54,10 €
    54,10 €
    1 * 24 ST
       54,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3026
  • 216-3027
    Plastic box, 6 compartments, compartment size 26×26 mm
    Plastic box, 6 compartments, compartment size 26×26 mm
    90×60×20 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 54,10 €
    54,10 €
    1 * 24 ST
       54,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3027
  • 216-3028
    Plastic box, 1 compartment, compartment size 121×72 mm
    Plastic box, 1 compartment, compartment size 121×72 mm
    130×90×25 mm
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 43,30 €
    43,30 €
    1 * 16 ST
       43,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3028
  • 216-3029
    Plastic box, 12 compartments, compartment size 29×26 mm
    Plastic box, 12 compartments, compartment size 29×26 mm
    130×90×25 mm
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 46,30 €
    46,30 €
    1 * 16 ST
       46,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3029
  • 216-3030
    Plastic box, 1 compartment, compartment size 171×118 mm
    Plastic box, 1 compartment, compartment size 171×118 mm
    190×130×30 mm
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 62,20 €
    62,20 €
    1 * 16 ST
       62,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3030
  • 216-3031
    Plastic box, 12 compartments, compartment size 40×38 mm
    Plastic box, 12 compartments, compartment size 40×38 mm
    190×130×30 mm
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 74,70 €
    74,70 €
    1 * 16 ST
       74,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3031
  • 216-3032
    Plastic box, 1 compartment, compartment size 250×153 mm
    Plastic box, 1 compartment, compartment size 250×153 mm
    260×180×40 mm
    Log in for availability
    Log in for availability
    1 * 14 ST
    1 * 14 ST - 105,00 €
    105,00 €
    1 * 14 ST
       105,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3032
  • 216-3033
    Plastic box, 24 compartments, compartment size 39×38 mm
    Plastic box, 24 compartments, compartment size 39×38 mm
    260×180×40 mm
    Log in for availability
    Log in for availability
    1 * 14 ST
    1 * 14 ST - 121,00 €
    121,00 €
    1 * 14 ST
       121,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3033
  • 216-3034
    Plastic box, 1 compartment, compartment size 351×239 mm
    Plastic box, 1 compartment, compartment size 351×239 mm
    360×260×50 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 89,10 €
    89,10 €
    1 * 6 ST
       89,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3034
  • 216-3035
    Plastic box, 28 compartments, compartment size 59×45 mm
    Plastic box, 28 compartments, compartment size 59×45 mm
    360×260×50 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 102,00 €
    102,00 €
    1 * 6 ST
       102,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    216-3035