ContactUsLinkComponent
HAEMAFlex™ 12/16 Swinging Bucket Rotors
HAEMAFlex™ 12/16 Swinging Bucket Rotors
Supplier:  Thermo Scientific
Certificates
undefined
HAEMAFlex™ 12/16 Swinging Bucket Rotors
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • HERA75003862
  • HERA75003882
  • Capacity:
  • 12× 500 ml blood bags (12× 1000 ml), 312× 5/7 ml blood tubes (oval buckets required)
  • 16× 500 ml blood bags (16× 1000 ml), 416× 5/7 ml blood tubes (oval buckets required)
  • Rotor type:
  • Swinging bucket rotor
  • Swinging bucket rotor
  • Max. RCF:
  • 7187×g
  • 5374×g
  • Max. speed:
  • 4700 min⁻¹
  • 3900 min⁻¹