Order Entry
Belgium
ContactUsLinkComponent
Orion™ Storage Sleeve and Base
Orion™ Storage Sleeve and Base
Catalog # ORIO810017
Supplier:  Thermo Orion
undefined
Orion™ Storage Sleeve and Base
Catalog # ORIO810017
Supplier:  Thermo Orion

Specifications

  • For:
    pH Electrode
  • Type:
    Storage sleeve, base
  • Description:
    Orion storage sleeve and base
  • Pk:
    1

Specifications

About this item

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ electrode storage sleeve and base.

  • It is for use with pH electrodes that are 120 mm or less in length that have a diameter of 12 mm
  • Sleeve protects your electrode from breakage when not in use and will not tip, due to the weighted removable base
  • As an additional convenience, the pH bulb stays conditioned while in storage so that it is ready to use when removed from the sleeve

Limited warranty period: Subject to all warranty requirements and exclusions set out in our terms and conditions of sale, this product is warranted from the date we ship the product and for 90 days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilise the products for internal research purposes only does not apply to the products on the web pages associated with this statement.