pH Sensor, Memosens CPS61E
About this item
Memosens 2.0 glass sensor for the food and beverage industry and for bioreactors in biotech industry.
- Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
- The sensor's exceptional accuracy and reproducibility help you to keep the pH value in the optimum range for a maximized product yield
- A special glass membrane and reference system makes the sensor CIP/SIP and autoclaving resistant (up to 140 °C/284 °F) offering maximum long-term stability
- The sensor protects your product quality with its certified biocompatibility according to USP class VI, FDA compliance of wetted parts, VO 1935/2004 compliance, no cytotoxicity, bioreactivity or animal-based materials
- Maximum process safety through non-contact, inductive signal transmission
- Reduced operating costs thanks to minimized process downtime and extended sensor lifetime
Keep the pH value in the optimum range and maximise your product yield with Memosens CPS61E. Its rugged design and long-term stability ensure extremely accurate and reproducible measured values even after CIP/SIP or autoclaving. CPS61E features Memosens 2.0 digital technology, offering extended storage of calibration and process data for predictive maintenance. The sensor resists moisture and enables pre-calibration in the laboratory. This enhances the process integrity and increases the process uptime.
Memosens CPS61E measures pH in: Biotech and pharma industries; bioreactors/fermenters: process control in enzyme production; bioreactors/fermenters: control of culture growth; food and beverage; CIP/SIP applications.
A pharma certificate of compliance with sensor serial number specified is available.
4 Options Available Below
Product Details & Documents
Memosens 2.0 glass sensor for the food and beverage industry and for bioreactors in biotech industry.
- Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
- The sensor's exceptional accuracy and reproducibility help you to keep the pH value in the optimum range for a maximized product yield
- A special glass membrane and reference system makes the sensor CIP/SIP and autoclaving resistant (up to 140 °C/284 °F) offering maximum long-term stability
- The sensor protects your product quality with its certified biocompatibility according to USP class VI, FDA compliance of wetted parts, VO 1935/2004 compliance, no cytotoxicity, bioreactivity or animal-based materials
- Maximum process safety through non-contact, inductive signal transmission
- Reduced operating costs thanks to minimized process downtime and extended sensor lifetime
Keep the pH value in the optimum range and maximise your product yield with Memosens CPS61E. Its rugged design and long-term stability ensure extremely accurate and reproducible measured values even after CIP/SIP or autoclaving. CPS61E features Memosens 2.0 digital technology, offering extended storage of calibration and process data for predictive maintenance. The sensor resists moisture and enables pre-calibration in the laboratory. This enhances the process integrity and increases the process uptime.
Memosens CPS61E measures pH in: Biotech and pharma industries; bioreactors/fermenters: process control in enzyme production; bioreactors/fermenters: control of culture growth; food and beverage; CIP/SIP applications.
A pharma certificate of compliance with sensor serial number specified is available.
Specifications for Product Family
Specifications
- Catalog No:
- 662-2462
- 662-2463
- ENDRCPS61E-1061/0
- ENDRCPS61E-10R4/0
- Range:
- 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)
- 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)
- 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)
- 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)
- Reference system:
- Pressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl
- Pressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl
- Pressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl
- Pressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl
- Sensor type:
- Hygienic version, zero point pH 7,0, temperature sensor NTC 30k
- Hygienic version, zero point pH 7,0, temperature sensor NTC 30k
- Hygienic version, zero point pH 7,0, temperature sensor NTC 30k
- Hygienic version, zero point pH 7,0, temperature sensor NTC 30k
Specifications
Product Family Options
Nonhazardous area approval
Product Information
- AvailabilityPrice
- 662-2462Memosens CPS61E pH sensor, 120 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EN 61326-1; EN 61326-2-3; EN IEC 63000; UKCA; Nonhazardous area approvalMemosens CPS61E pH sensor, 120 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EN 61326-1; EN 61326-2-3; EN IEC 63000; UKCA; Nonhazardous area approvalLog in for availability1 * 1 ST809,00 €Specifications:
More
Specifications
Cat. No.662-2462Range0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)Reference systemPressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgClSensor typeHygienic version, zero point pH 7,0, temperature sensor NTC 30k - 662-2463Memosens CPS61E pH sensor, 225 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EC_00831_03.20; Nonhazardous area approvalMemosens CPS61E pH sensor, 225 mm, 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs), sterilisable up to 140 °C; EC_00831_03.20; Nonhazardous area approvalLog in for availability1 * 1 ST927,00 €Specifications:
More
Specifications
Cat. No.662-2463Range0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)Reference systemPressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgClSensor typeHygienic version, zero point pH 7,0, temperature sensor NTC 30k
ATEX approval
Product Information
- AvailabilityPrice
- ENDRCPS61E-1061/0pH sensor (CPS61E-BA6NTP2); ATEX II 1G Ex ia IIC T3/T4/T6 Ga; process connect. Pg 13.5; hygienic version, zero point pH 7,0, temperature sensor NTC 30k; ref. system pressurized, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl; 120 mm shaft; 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilizable up to 140 °CpH sensor (CPS61E-BA6NTP2); ATEX II 1G Ex ia IIC T3/T4/T6 Ga; process connect. Pg 13.5; hygienic version, zero point pH 7,0, temperature sensor NTC 30k; ref. system pressurized, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl; 120 mm shaft; 0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilizable up to 140 °CLog in for availability1 * 1 ST824,00 €Specifications:
More
Specifications
Cat. No.ENDRCPS61E-1061/0Range0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)Reference systemPressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgClSensor typeHygienic version, zero point pH 7,0, temperature sensor NTC 30k - ENDRCPS61E-10R4/0pH sensor (CPS61E-BA6NTP4); ATEX II 1G Ex ia IIC T3/T4/T6 Ga; process connect. Pg 13.5; hygienic version, zero point pH 7,0, temperature sensor NTC 30k; ref. system pressurized, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl; 225 mm shaft;0 -1 4 pH, 0...100 °C, 0,8...7 bar (abs), sterilizable up to 140 °CpH sensor (CPS61E-BA6NTP4); ATEX II 1G Ex ia IIC T3/T4/T6 Ga; process connect. Pg 13.5; hygienic version, zero point pH 7,0, temperature sensor NTC 30k; ref. system pressurized, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgCl; 225 mm shaft;0 -1 4 pH, 0...100 °C, 0,8...7 bar (abs), sterilizable up to 140 °CLog in for availability1 * 1 ST960,00 €Specifications:
More
Specifications
Cat. No.ENDRCPS61E-10R4/0Range0 - 14 pH, 0...100 °C, 0,8...7 bar (abs) , sterilisable up to 140 °C (application range)Reference systemPressurised, 1 ceramic junction, ion trap, 3 M KCl, Ag/AgClSensor typeHygienic version, zero point pH 7,0, temperature sensor NTC 30k
Compatible fermenter set
Product Information
- Catalog #ConnectionDescriptionAvailabilityPrice
- Memosens input (T82 and KSG8 standard adapter out)CYM17 Memosens-analog converter set 2 for lab fermenters with 225 mm Memosens 2,0 sensorsLog in for availability1 * 1 ST4 550,00 €