ContactUsLinkComponent
Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
undefined
Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
  • Catalog No:
  • STUA51600-94
  • STUA51600-99
  • STUA51600-91
  • STUA51600-98
  • STUA51600-97
  • STUA51600-96
  • STUA51600-95
  • STUA51600-93
  • STUA51600-92
  • STUA51600-06
  • Speed range:
  • 6 - 60 min⁻¹
  • W×D×H:
  • 650×250×250 mm
  • Electrical:
  • 230 VAC, 50 Hz
  • Max. load:
  • 3 kg
  • Timer:
  • 10 - 60 min
Frequently Bought Together