Order Entry
Belgium
ContactUsLinkComponent
µSIL-WAX Capillaries with Windows, Agilent Technologies
µSIL-WAX Capillaries with Windows, Agilent Technologies
Catalog # HEWL197-7202
Supplier:  Agilent
µSIL-WAX Capillaries with Windows, Agilent Technologies
Catalog # HEWL197-7202
Supplier:  Agilent

Specifications

  • Description:
    μSIL-WAX capillaries with windows, ID 100 μm, film thickness 0,1 μm
  • Seq:
    4
  • Total length:
    1000 mm
  • Pk:
    1
  • Usable length:
    750 mm

Specifications

About this item

µSIL-WAX capillaries with windows, featuring a modified, polyethylene oxide, hydrophilic coating.

  • Ideal for CE/MS
  • Ideal for protein and peptide separations from pH 2 to 5

µSIL-WAX capillaries feature a modified, polyethylene oxide, hydrophilic coating. The coating effectively masks active silanol sites. The highly stable coating and near-zero EOF of µSIL-WAX makes the capillary ideal for CE-MS, and protein and peptide separations.