ContactUsLinkComponent
Dispersing Tools for OV 625 Digital and OV 725 Digital
Dispersing Tools for OV 625 Digital and OV 725 Digital
Supplier:  VELP SCIENTIFIC
Certificates
undefined
Dispersing Tools for OV 625 Digital and OV 725 Digital
Supplier:  VELP SCIENTIFIC
Specifications
  • Catalog No:
  • VELPA00000472
  • VELPA00000469
  • VELPA00000464
  • VELPA00000463
  • VELPA00000479
  • VELPA00000480
  • VELPA00000462
  • VELPA00000478
  • VELPA00000473
  • Gap between rotor and stator:
  • 8 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • 0,5 mm
  • Circumferential speed:
  • 18,8 m/s
  • 23,6 m/s
  • 19,6 m/s
  • 19,6 m/s
  • 23,6 m/s
  • 23,6 m/s
  • 19,6 m/s
  • 19,6 m/s
  • 4,2 m/s
  • Ultimate fineness:
  • -
  • 1 - 5 µm (emulsion)
    5 - 25 µm (suspensions)
  • 1 - 7 µm (emulsion)
    5 - 40 µm (suspensions)
  • 1 - 7 µm (emulsion)
    5 - 40 µm (suspensions)
  • 1 - 10 µm (emulsion)
    10 - 50 µm (suspensions)
  • 1 - 5 µm (emulsion)
    5 - 25 µm (suspensions)
  • 1 - 7 µm (emulsion)
    5 - 40 µm (suspensions)
  • 1 - 10 µm (emulsion)
    10 - 50 µm (suspensions)
  • 1 - 10 µm (emulsion)
    10 - 50 µm (suspensions)