ContactUsLinkComponent
Module for AiroSensor 20-20-41, Magnetic Door Contact Set
Module for AiroSensor 20-20-41, Magnetic Door Contact Set
Catalog # SENA0096647495187
Supplier:  SenseAnywhere
undefined
Module for AiroSensor 20-20-41, Magnetic Door Contact Set
Catalog # SENA0096647495187
Supplier:  SenseAnywhere
Specifications
  • Description:
    Module for AiroSensor 20-20-41, magnetic door contact set, 5 m cable length, magnet size 50L×14W×12D mm, 100 g weight (door contact set only; AiroSensor 20-20-41 not included)
  • Pk:
    1
Specifications
About this item
The SenseAnywhere AiroSensor 20-20-41 with Magnetic Door Contact Set is the ultimate solution to your refrigeration and freezer challenges!
  • Upgrade your systems today!
We all know the frustration of discovering that your fridge or freezer has failed due to an open door. With the AiroSensor 20-20-41 combined with the Magnetic Door Contact Set (both sold separately), you can say goodbye to this common problem.
With these two products, which should both be ordered; you can program alarms for open or improperly closed doors. This innovative solution monitors door frequency and duration on refrigerators and freezers to prevent potential disasters caused by open doors.
The connecter with cable can be easily shortened and the magnetic switch set with terminal screws can easily mount the cable, and they are provided with two double-sided nano-tape pads for easy mounting even in sub-zero temperatures. Please note that the black jack is removed from the 20-20-41 products.
Experience has shown that most alarms and excursions occur due to improperly closed doors. But with the AiroSensor 20-20-41 and Magnetic Door Contact Set (both sold separately), you can rest easy knowing that you are protected against these potential disasters.
Enjoy the peace of mind that comes with knowing your refrigeration and freezer systems are operating at peak performance. Don't wait any longer - upgrade your systems today and join the thousands of satisfied customers who have already experienced the benefits of SenseAnywhere technology. The Magnetic Door Contact Set is a reliable and easy-to-install accessory for use with the AiroSensor 20-20-41. With a 5-metre cable and flexible attachment options, this accessory is the perfect solution for monitoring fridge doors. This can be easily attached to the fridge using tape or screws, depending on your preference. An alcohol wipe is included to clean surfaces before installation, ensuring optimal adhesion.
Ordering information: Please note that this module is only suitable for use with the AiroSensor 20-20-41. The AiroSensor 20-20-41 and the magnetic door contact set are not supplied together and must be ordered separately. Contact your Avantor representative for more information.