Rapid Adapter Auxiliary V14
Catalogus # OXNTEXP-RAA114
Leverancier: Oxford Nanopore Technologies
CAS-nummer:
Specificaties
- Pk:1 KIT
- Description:Rapid Adapter Auxiliary V14
- Compatible with:R10.4.1 flow cells
- No. of reactions:6
Specificaties
Over dit artikel
Additional rapid adapter and adapter buffer to enable customers to maximise the use of rapid barcoding kits 24 and 96 V14 (SQK-RBK114.24 and SQK-RBK114.96).
The rapid adapter auxiliary V14 (EXP-RAA114) expansion contains additional rapid adapter and adapter buffer for another six reactions. This is to enable customers to use any unused barcodes in their rapid barcoding kits V14.
To use this kit, ensure to purchase the sequencing auxiliary vials V14 (EXP-AUX003) to provide the flow cell priming reagents.
Levering: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.