Orders LinkContactUsLinkComponent
Powder Funnels, PP
Powder Funnels, PP
Supplier:   KARTELL
Powder Funnels, PP
Supplier:  KARTELL
Certificates
Powder Funnels, PP
Powder Funnels, PP
6 Options Available Below
Product Details & Documents
60° angle, autoclavable funnels.
  • Large bore avoids bridging of powders during transfer
  • Ribbed outside to avoid pressure lock
  • Excellent chemical resistance
Can be used with liquids.
Certifications: The raw materials used comply with food-contact EU regulation 1935/2004, 10/2011, US Food and Drug Administration and REACH/ROHS regulation.
Product Family Options
Product Information
  • Funnel O-Ø
    Length
    Stem O-Ø
    Material
    Stem length
    Availability
    Price
  • KART0017100
    45 ml capacity
    45 ml capacity
    60 mm
    57,0 mm
    15 mm
    PP
    18,0 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 49,60 €
    49,60 €
    1 * 20 ST
       49,60 €
  • KART0016800
    200 ml capacity
    200 ml capacity
    100 mm
    88,5 mm
    25 mm
    PP
    24,8 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 114,00 €
    114,00 €
    1 * 20 ST
       114,00 €
  • KART0016900
    360 ml capacity
    360 ml capacity
    120 mm
    106,3 mm
    30 mm
    PP
    27,1 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 71,70 €
    71,70 €
    1 * 10 ST
       71,70 €
  • KART0016700
    730 ml capacity
    730 ml capacity
    80 mm
    79,8 mm
    36 mm
    PP
    22,2 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 109,00 €
    109,00 €
    1 * 20 ST
       109,00 €
  • KART0017000
    730 ml capacity
    730 ml capacity
    150 mm
    142,0 mm
    36 mm
    PP
    39,9 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 88,50 €
    88,50 €
    1 * 10 ST
       88,50 €
  • 221-1536
    1290 ml capacity
    1290 ml capacity
    180 mm
    166,0 mm
    43 mm
    PP
    48,8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,50 €
    56,50 €
    1 * 5 ST
       56,50 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.