ContactUsLinkComponent
Culti-Loops™ Paenibacillus polymyxa ATCC™ 7070™
Culti-Loops™ Paenibacillus polymyxa ATCC™ 7070™
Catalog # OXOIR4604600
CAS Number:  
undefined
Culti-Loops™ Paenibacillus polymyxa ATCC™ 7070™
Catalog # OXOIR4604600
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Paenibacillus polymyxa ATCC™ 7070™
  • Environmentally Preferable:
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.