ContactUsLinkComponent
Culti-Loops™ Haemophilus influenzae ATCC™ 19418™
Culti-Loops™ Haemophilus influenzae ATCC™ 19418™
N° de catalogue OXOIR4603851
Numéro CAS:  
undefined
Culti-Loops™ Haemophilus influenzae ATCC™ 19418™
N° de catalogue OXOIR4603851
Numéro CAS:  
Spécifications
  • Pk:
    5
  • Description:
    Haemophilus influenzae ATCC™ 19418™
  • Environmentally Preferable:
Spécifications
À propos de cet article
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Emballage: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.