B.O.D. Bottle Rack, Poxygrid®
Poxygrid® B.O.D. bottle rack, W×D×H:24,8×33,8×18,4 cm
Catalog # BELAF169690012
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
Racks have 12 places (3×4 rows) for 300 ml Biological Oxygen Demand bottles.
- Epoxy-coated steel wire construction
- Chemical- and corrosion-resistant
- Stackable for storage and refrigeration
- Autoclavable
Made from epoxy-coated steel wire, they are stackable for storage or refrigeration, allowing 16,7 cm between racks. Chemical- and corrosion-resistant, they are autoclavable at 121 °C.
Specifications
- Description:Poxygrid® B.O.D. bottle rack, W×D×H:24,8×33,8×18,4 cm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



