Orders LinkContactUsLinkComponent
Separating Funnels, Squibb
Separating Funnels, Squibb
Supplier:   Kavalier
Separating Funnels, Squibb
Supplier:  Kavalier
Certificates
Separating Funnels, Squibb
Separating Funnels, Squibb
5 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 50 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 299,00 €
    299,00 €
    1 * 2 ST
       299,00 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 700,00 €
    700,00 €
    1 * 2 ST
       700,00 €