Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium polyphosphate (Graham's salt)
Sodium polyphosphate (Graham's salt), glassy FCC
Sodium polyphosphate (Graham's salt), glassy FCC
Sodium polyphosphate (Graham's salt), glassy FCC
SPCMSO169-500GM
:
Spectrum Chemical
:
Sodium polyphosphate (Graham's salt), glassy FCC
SPCMSO169-500GM
:
Spectrum Chemical
:
Pack type:
Poly bottle
Pk:
500 g
Seq:
3645
Assay:
60% Min.
:
:
Ambient
:
256-779-4
:
50813-16-6
:
(NaO₃P)n
:
MFCD00134118