Ammonia solution 27 - 30% FCC
: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650
:0,900
:35.05 g/mol
:2672
:8 III
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
- Assay (NH₃, By Wt.):27.0 - 30.0%
- Lead (Pb):max. 0.5 mg/kg
- Nonvolatile Residue:max. 0.02 %
- Readily Oxidizable Substances:To Pass Test
- Identification:To Pass Test
Frequently Bought Together