Order Entry
Belgium
ContactUsLinkComponent
Lactic acid 85% ≥85% FCC
Corrosive
Lactic acid 85% ≥85% FCC
Supplier:  Spectrum Chemical
Certificates
Corrosive
Lactic acid 85% ≥85% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (C₃H₆O₃):
    81 - 89%
  • Chloride (Cl):
    max. 0.1%
  • Citric, Oxalic, Phosphoric and Tartaric Acid:
    To Pass Test
  • Cyanide:
    max. 5 mg/kg
  • Iron (Fe):
    max. 10 mg/kg

Specification Test Results

Frequently Bought Together