Order Entry
Belgium
ContactUsLinkComponent
Hydroxypropyl methylcellulose (Hypromellose), 3,550 mPa.s USP Substitution Type 2208
Hydroxypropyl methylcellulose (Hypromellose), 3,550 mPa.s USP Substitution Type 2208
Supplier:  Spectrum Chemical
Certificates
Hydroxypropyl methylcellulose (Hypromellose), 3,550 mPa.s USP Substitution Type 2208
Supplier:  Spectrum Chemical

Specification Test Results

  • Viscosity of a 2% Aqueous Solution @ 20D °C:
    2663 - 4970 mPa·s
  • Assay (Methoxy Groups):
    19.0 - 24.0%
  • Assay (Hydroxypropoxy Groups):
    4.0 - 12.0%
  • pH:
    5.0 - 8.0
  • Loss On Drying:
    max. 5.0%

Specification Test Results

Frequently Bought Together